Product Details

SNP ID
rs11795728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141847241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACAAGCACTTGTTACCCTCTTTC[A/G]CGTTATTCAACCAAAAGAGTTGTAG
Phenotype
MIM: 300469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEC3 PubMed Links

Gene Details

Gene
MAGEC3
Gene Name
MAGE family member C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138702.1 Intron NP_619647.1
NM_177456.2 Intron NP_803251.1
XM_011531267.2 Intron XP_011529569.1
XM_017029265.1 Intron XP_016884754.1

View Full Product Details