Product Details

SNP ID
rs2285177
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:37809887 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCACTGACGCTCCTTCTGTAGCTGT[C/T]GAATTTGGTGCCTCCTCAATGGTGA
Phenotype
MIM: 607422 MIM: 142708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCAT PubMed Links

Gene Details

Gene
GCAT
Gene Name
glycine C-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171690.1 Intron NP_001165161.1
NM_014291.3 Intron NP_055106.1
XM_005261409.3 Intron XP_005261466.1
XM_005261410.3 Intron XP_005261467.1
XM_005261411.1 Intron XP_005261468.1
XM_011530023.1 Intron XP_011528325.1
XM_017028674.1 Intron XP_016884163.1
XM_017028675.1 Intron XP_016884164.1
XM_017028676.1 Intron XP_016884165.1
XM_017028677.1 Intron XP_016884166.1
Gene
H1F0
Gene Name
H1 histone family member 0
There are no transcripts associated with this gene.

View Full Product Details