Product Details

SNP ID
rs34177663
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:7773168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAAGGACAGTAGAGCTTACCT[A/G]TCGTGGGATCAGATACTTCAGGGAT
Phenotype
MIM: 611501 MIM: 603657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMTA1 PubMed Links

Gene Details

Gene
CAMTA1
Gene Name
calmodulin binding transcription activator 1
There are no transcripts associated with this gene.

Gene
VAMP3
Gene Name
vesicle associated membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004781.3 Intron NP_004772.1
XM_017002841.1 Intron XP_016858330.1

View Full Product Details