Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001289861.1 | Intron | NP_001276790.1 | |||
NM_001289862.1 | Intron | NP_001276791.1 | |||
NM_001289863.1 | Intron | NP_001276792.1 | |||
NM_001289864.1 | Intron | NP_001276793.1 | |||
NM_016831.2 | Intron | NP_058515.1 | |||
XM_005263524.3 | Intron | XP_005263581.1 | |||
XM_011542385.2 | Intron | XP_011540687.1 | |||
XM_011542390.2 | Intron | XP_011540692.1 | |||
XM_017002723.1 | Intron | XP_016858212.1 | |||
XM_017002724.1 | Intron | XP_016858213.1 | |||
XM_017002725.1 | Intron | XP_016858214.1 | |||
XM_017002726.1 | Intron | XP_016858215.1 | |||
XM_017002727.1 | Intron | XP_016858216.1 | |||
XM_017002728.1 | Intron | XP_016858217.1 | |||
XM_017002729.1 | Intron | XP_016858218.1 | |||
XM_017002730.1 | Intron | XP_016858219.1 | |||
XM_017002731.1 | Intron | XP_016858220.1 | |||
XM_017002732.1 | Intron | XP_016858221.1 | |||
XM_017002733.1 | Intron | XP_016858222.1 | |||
XM_017002734.1 | Intron | XP_016858223.1 | |||
XM_017002735.1 | Intron | XP_016858224.1 | |||
XM_017002736.1 | Intron | XP_016858225.1 | |||
XM_017002737.1 | Intron | XP_016858226.1 | |||
XM_017002738.1 | Intron | XP_016858227.1 |