Product Details

SNP ID
rs228651
Assay Type
Validated
NCBI dbSNP Submissions
75
Location
Chr.1:7851039 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AGATGAACACTAGAAAAACTTCCAA[C/T]GCTGAGTTCATGACGTGTCATCAGT
Phenotype
MIM: 603427 MIM: 604097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PER3 PubMed Links

Gene Details

Gene
PER3
Gene Name
period circadian clock 3
There are no transcripts associated with this gene.

Gene
UTS2
Gene Name
urotensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006786.3 Intron NP_006777.1
NM_021995.2 Intron NP_068835.1
XM_011540537.2 Intron XP_011538839.1
XM_011540538.1 Intron XP_011538840.1
XM_017000116.1 Intron XP_016855605.1
XM_017000117.1 Intron XP_016855606.1
XM_017000118.1 Intron XP_016855607.1
XM_017000119.1 Intron XP_016855608.1

View Full Product Details