Product Details

SNP ID
rs514552
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138090377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAACAAATGATAAAATGGGAAGG[A/G]GGGCAAGGGGAAAATAGAGAGACTG
Phenotype
MIM: 609301
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PERP PubMed Links

Gene Details

Gene
PERP
Gene Name
PERP, TP53 apoptosis effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022121.4 2430 UTR 3 NP_071404.2

View Full Product Details