Product Details

SNP ID
rs8155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138090933 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAAAAATCATAATGACCTATCC[A/G]ATGCATCATATATATGCTATTCAGA
Phenotype
MIM: 609301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PERP PubMed Links

Gene Details

Gene
PERP
Gene Name
PERP, TP53 apoptosis effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022121.4 1874 UTR 3 NP_071404.2

View Full Product Details