Product Details

SNP ID
rs17003556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43206845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACGCATCCCTGGTTTCTGCC[C/T]AAGCAGCTTGAGGAGGAAATCAGGA
Phenotype
MIM: 611746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105373051 PubMed Links

Gene Details

Gene
LOC105373051
Gene Name
uncharacterized LOC105373051
There are no transcripts associated with this gene.

Gene
SCUBE1
Gene Name
signal peptide, CUB domain and EGF like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173050.3 Intron NP_766638.2
XM_005261752.1 Intron XP_005261809.1
XM_011530389.2 Intron XP_011528691.1
XM_011530390.2 Intron XP_011528692.1

View Full Product Details