Product Details

SNP ID
rs2143747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46137682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCATTAGGTTTGTTACTCTGTC[C/T]TTTGGAGAGGTGGTTTTGTTACTCT
Phenotype
MIM: 607293 MIM: 617000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIC5 PubMed Links

Gene Details

Gene
CLIC5
Gene Name
chloride intracellular channel 5
There are no transcripts associated with this gene.

Gene
ENPP4
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014936.4 Intron NP_055751.1

View Full Product Details