Product Details

SNP ID
rs34213429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41103222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGGGTAGGCAGGTGTGATGGG[A/G]GTAGTGATGTTACATTTGGGGCATT
Phenotype
MIM: 602700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EP300 PubMed Links

Gene Details

Gene
EP300
Gene Name
E1A binding protein p300
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001429.3 Intron NP_001420.2
XM_006724165.3 Intron XP_006724228.1

View Full Product Details