Product Details

SNP ID
rs9257694
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:29306709 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGACTATGGTCAATTTGACTTCAA[C/T]GAGTGGATTCCTTCTTATGGGGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14J1 PubMed Links

Gene Details

Gene
OR14J1
Gene Name
olfactory receptor family 14 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030946.1 20 Missense Mutation ACG,ATG T7M NP_112208.1

View Full Product Details