Product Details
- SNP ID
-
rs6048797
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23441984 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CCAATGCAGACTGTCCCACAGGCTG[A/G]CTCAGTGAGGCACTTCTACCACAAG
- Phenotype
-
MIM: 609731
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST11
PubMed Links
Gene Details
- Gene
- CST11
- Gene Name
- cystatin 11
There are no transcripts associated with this gene.
- Gene
- CSTL1
- Gene Name
- cystatin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138283.1 |
|
Intron |
|
|
NP_612140.1 |
View Full Product Details