Product Details

SNP ID
rs4815198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23362854 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTGTTGATTGAGAAATCAGGTC[C/T]CCTCGAGGGCCCTGTTGGCAGAGGT
Phenotype
MIM: 613842 MIM: 605811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZF1 PubMed Links

Gene Details

Gene
GZF1
Gene Name
GDNF inducible zinc finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317012.1 Intron NP_001303941.1
NM_001317019.1 Intron NP_001303948.1
NM_022482.4 Intron NP_071927.1
XM_005260800.2 Intron XP_005260857.1
XM_005260802.3 Intron XP_005260859.1
XM_011529321.2 Intron XP_011527623.1
XM_011529322.1 Intron XP_011527624.1
XM_017028017.1 Intron XP_016883506.1
XM_017028018.1 Intron XP_016883507.1
XM_017028019.1 Intron XP_016883508.1
Gene
LINC01431
Gene Name
long intergenic non-protein coding RNA 1431
There are no transcripts associated with this gene.

Gene
NXT1
Gene Name
nuclear transport factor 2 like export factor 1
There are no transcripts associated with this gene.

View Full Product Details