Product Details

SNP ID
rs393427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108168396 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGGTGTGGAGCCCCTGGGGCCCT[A/T]GAAAGGTGAGGCCTCAGAGGCGAGG
Phenotype
MIM: 603849
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NR2E1 PubMed Links

Gene Details

Gene
NR2E1
Gene Name
nuclear receptor subfamily 2 group E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286102.1 Intron NP_001273031.1
NM_003269.4 Intron NP_003260.1
Gene
OSTM1-AS1
Gene Name
OSTM1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details