Product Details

SNP ID
rs2273145
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:49223054 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGGCTATTTTTCGTTGTTAGGGCC[C/T]ACTCTTTACTCTCACCACGACCCCA
Phenotype
MIM: 616621 MIM: 601716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX27 PubMed Links

Gene Details

Gene
DDX27
Gene Name
DEAD-box helicase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017895.7 Intron NP_060365.7
XM_011528888.2 Intron XP_011527190.1
Gene
STAU1
Gene Name
staufen double-stranded RNA binding protein 1
There are no transcripts associated with this gene.

View Full Product Details