Product Details

SNP ID
rs12008596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:105832491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCAGGCCTGTTGCTCAAAAAC[C/T]GGATTACTGTCCTTAGGCTGATCCT
Phenotype
MIM: 300791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRK PubMed Links

Gene Details

Gene
NRK
Gene Name
Nik related kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198465.3 Intron NP_940867.2
XM_006724632.3 Intron XP_006724695.1
XM_006724633.3 Intron XP_006724696.1
XM_006724634.3 Intron XP_006724697.1
XM_011530887.2 Intron XP_011529189.1

View Full Product Details