Product Details

SNP ID
rs12007249
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:37690999 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAGTTTTCCTAACCAACAGTGGCA[A/G]GAGGTCTTGGGGCCTTTTACAAATG
Phenotype
MIM: 314850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LANCL3 PubMed Links

Gene Details

Gene
LANCL3
Gene Name
LanC like 3
There are no transcripts associated with this gene.

Gene
XK
Gene Name
X-linked Kx blood group
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011543978.2 Intron XP_011542280.1

View Full Product Details