Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170701.1 | Intron | NP_001164172.1 | |||
NM_001170702.1 | Intron | NP_001164173.1 | |||
NM_001170703.1 | Intron | NP_001164174.1 | |||
NM_001170704.1 | Intron | NP_001164175.1 | |||
NM_018388.3 | Intron | NP_060858.2 | |||
NM_133486.2 | Intron | NP_597846.1 | |||
XM_005262431.3 | Intron | XP_005262488.1 | |||
XM_005262434.3 | Intron | XP_005262491.1 | |||
XM_005262436.3 | Intron | XP_005262493.1 | |||
XM_005262437.3 | Intron | XP_005262494.1 | |||
XM_005262438.3 | Intron | XP_005262495.1 | |||
XM_005262439.3 | Intron | XP_005262496.1 | |||
XM_005262440.3 | Intron | XP_005262497.1 | |||
XM_005262441.3 | Intron | XP_005262498.1 | |||
XM_005262442.3 | Intron | XP_005262499.1 | |||
XM_011531364.2 | Intron | XP_011529666.1 | |||
XM_017029632.1 | Intron | XP_016885121.1 | |||
XM_017029633.1 | Intron | XP_016885122.1 | |||
XM_017029634.1 | Intron | XP_016885123.1 | |||
XM_017029635.1 | Intron | XP_016885124.1 | |||
XM_017029636.1 | Intron | XP_016885125.1 | |||
XM_017029637.1 | Intron | XP_016885126.1 | |||
XM_017029638.1 | Intron | XP_016885127.1 | |||
XM_017029639.1 | Intron | XP_016885128.1 |