Product Details
- SNP ID
-
rs14034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31619420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACGGGATAGGTTGAGGGGCATGA[C/T]GGGGGCTCTCGCCACCTCTTGTCTG
- Phenotype
-
MIM: 612770
MIM: 612765
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7109
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142338374] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR7109
- Gene Name
- microRNA 7109
There are no transcripts associated with this gene.
- Gene
- PISD
- Gene Name
- phosphatidylserine decarboxylase
- Gene
- SFI1
- Gene Name
- SFI1 centrin binding protein
View Full Product Details