Product Details
- SNP ID
-
rs658591
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:28283743 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTATATACTGTGCTTCATAGTTCAC[G/T]TGTTTTATCAACCTTCATAGTTATA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT18P55
PubMed Links
Gene Details
- Gene
- KRT18P55
- Gene Name
- keratin 18 pseudogene 55
There are no transcripts associated with this gene.
View Full Product Details