Product Details

SNP ID
rs1050700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132892556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTATCCGCAGGAGGTGGGCGGCAC[C/T]CCATCTCACCAAGGTCCTGGCCGGG
Phenotype
MIM: 605284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPACA9 PubMed Links

Gene Details

Gene
SPACA9
Gene Name
sperm acrosome associated 9
There are no transcripts associated with this gene.

Gene
TSC1
Gene Name
tuberous sclerosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000368.4 7453 UTR 3 NP_000359.1
NM_001162426.1 7453 UTR 3 NP_001155898.1
NM_001162427.1 7453 UTR 3 NP_001155899.1
XM_005272211.1 7453 UTR 3 XP_005272268.1
XM_006717271.1 7453 UTR 3 XP_006717334.1
XM_011518979.2 7453 UTR 3 XP_011517281.1
XM_017015096.1 7453 UTR 3 XP_016870585.1
XM_017015097.1 7453 UTR 3 XP_016870586.1
XM_017015098.1 7453 UTR 3 XP_016870587.1
XM_017015099.1 7453 UTR 3 XP_016870588.1
XM_017015100.1 7453 UTR 3 XP_016870589.1
XM_017015101.1 7453 UTR 3 XP_016870590.1

View Full Product Details