Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134398.1 | 3097 | UTR 3 | NP_001127870.1 | ||
NM_003371.3 | 3097 | UTR 3 | NP_003362.2 | ||
XM_005272213.1 | 3097 | UTR 3 | XP_005272270.1 | ||
XM_017015108.1 | 3097 | UTR 3 | XP_016870597.1 | ||
XM_017015109.1 | 3097 | UTR 3 | XP_016870598.1 | ||
XM_017015110.1 | 3097 | UTR 3 | XP_016870599.1 | ||
XM_017015111.1 | 3097 | UTR 3 | XP_016870600.1 | ||
XM_017015112.1 | 3097 | UTR 3 | XP_016870601.1 | ||
XM_017015113.1 | 3097 | Intron | XP_016870602.1 |