Product Details
- SNP ID
-
rs7170454
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:90718536 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATGAGAGATCAGAAGATGAAGAAG[C/G]GGGTAGGCAAAAGTATTCTGTTTGG
- Phenotype
-
MIM: 604610
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BLM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115175743] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BLM
- Gene Name
- Bloom syndrome RecQ like helicase
- Gene
- CRTC3-AS1
- Gene Name
- CRTC3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details