Product Details

SNP ID
rs1233554
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:155806185 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCGGAAACGCAGTCATCGCCCAGCG[A/C]CCCTGCTCCTGGCCCTCAGCCTCCC
Phenotype
MIM: 600725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SHH PubMed Links

Gene Details

Gene
SHH
Gene Name
sonic hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000193.3 Intron NP_000184.1
NM_001310462.1 Intron NP_001297391.1
XM_011516479.2 Intron XP_011514781.1
XM_011516480.2 Intron XP_011514782.1

View Full Product Details