Product Details

SNP ID
rs9304802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57698170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGATGAATCTCAAAATAAATAC[A/G]TGACTATACCCAGGGATTAAAAAAG
Phenotype
MIM: 604085
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF154 PubMed Links

Gene Details

Gene
ZNF154
Gene Name
zinc finger protein 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085384.2 5019 UTR 3 NP_001078853.1
Gene
ZNF551
Gene Name
zinc finger protein 551
There are no transcripts associated with this gene.

View Full Product Details