Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308068.1 | 389 | UTR 5 | NP_001294997.1 | ||
NM_020912.1 | 389 | Intron | NP_065963.1 | ||
NM_032296.2 | 389 | UTR 5 | NP_115672.2 | ||
XM_006720962.3 | 389 | UTR 5 | XP_006721025.1 | ||
XM_011522701.2 | 389 | UTR 5 | XP_011521003.1 | ||
XM_011522702.2 | 389 | UTR 5 | XP_011521004.1 | ||
XM_011522703.2 | 389 | UTR 5 | XP_011521005.1 | ||
XM_011522704.2 | 389 | UTR 5 | XP_011521006.1 | ||
XM_011522705.2 | 389 | UTR 5 | XP_011521007.1 | ||
XM_011522707.2 | 389 | UTR 5 | XP_011521009.1 | ||
XM_017023772.1 | 389 | UTR 5 | XP_016879261.1 |