Product Details

SNP ID
rs34106761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7225506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCAACCCTGCAAAGGGCAGG[G/T]CTGTGGGTAACTGGAGGAGGAGGTC
Phenotype
MIM: 609575 MIM: 602887 MIM: 602151 MIM: 612910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACADVL PubMed Links

Gene Details

Gene
ACADVL
Gene Name
acyl-CoA dehydrogenase, very long chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000018.3 2198 Intron NP_000009.1
NM_001033859.2 2198 Intron NP_001029031.1
NM_001270447.1 2198 Intron NP_001257376.1
NM_001270448.1 2198 Intron NP_001257377.1
XM_006721516.2 2198 Intron XP_006721579.2
XM_011523829.1 2198 Intron XP_011522131.1
XM_011523830.1 2198 Intron XP_011522132.1
Gene
DLG4
Gene Name
discs large MAGUK scaffold protein 4
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004422.2 2198 UTR 3 NP_004413.1
XM_005256502.1 2198 UTR 3 XP_005256559.1
XM_017024298.1 2198 UTR 3 XP_016879787.1
Gene
MIR324
Gene Name
microRNA 324
There are no transcripts associated with this gene.

Gene
PHF23
Gene Name
PHD finger protein 23
There are no transcripts associated with this gene.

View Full Product Details