Product Details

SNP ID
rs11748773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132419069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTTACCCACAATTTTATCTGG[C/T]ATAGTGCCTGGCATGCAGCTGTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf56 PubMed Links

Gene Details

Gene
C5orf56
Gene Name
chromosome 5 open reading frame 56
There are no transcripts associated with this gene.

View Full Product Details