Product Details
- SNP ID
-
rs2301762
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3500977 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTGACCGTGCGCCGTCCAAGGGTC[C/G]ATTGGTTGCCATAGAGATCGTCGAG
- Phenotype
-
MIM: 616787
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C16orf90
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111497094] are located under a probe and SNP(s) [rs79384355] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C16orf90
- Gene Name
- chromosome 16 open reading frame 90
There are no transcripts associated with this gene.
- Gene
- CLUAP1
- Gene Name
- clusterin associated protein 1
View Full Product Details