Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024732.2 | 1583 | UTR 3 | NP_001019903.2 | ||
NM_016011.3 | 1583 | UTR 3 | NP_057095.3 | ||
XM_005245885.2 | 1583 | UTR 3 | XP_005245942.1 | ||
XM_005245887.2 | 1583 | Intron | XP_005245944.1 | ||
XM_011541539.1 | 1583 | Intron | XP_011539841.1 | ||
XM_011541540.1 | 1583 | Intron | XP_011539842.1 | ||
XM_011541541.1 | 1583 | Intron | XP_011539843.1 | ||
XM_011541543.1 | 1583 | Intron | XP_011539845.1 | ||
XM_011541545.1 | 1583 | Intron | XP_011539847.1 | ||
XM_011541546.1 | 1583 | Intron | XP_011539848.1 | ||
XM_011541547.1 | 1583 | Intron | XP_011539849.1 | ||
XM_011541549.1 | 1583 | Intron | XP_011539851.1 | ||
XM_011541550.1 | 1583 | Intron | XP_011539852.1 | ||
XM_011541553.1 | 1583 | Intron | XP_011539855.1 | ||
XM_011541554.1 | 1583 | Intron | XP_011539856.1 | ||
XM_017001411.1 | 1583 | Intron | XP_016856900.1 | ||
XM_017001412.1 | 1583 | Intron | XP_016856901.1 | ||
XM_017001413.1 | 1583 | Intron | XP_016856902.1 | ||
XM_017001414.1 | 1583 | Intron | XP_016856903.1 | ||
XM_017001415.1 | 1583 | Intron | XP_016856904.1 | ||
XM_017001416.1 | 1583 | Intron | XP_016856905.1 |