Product Details

SNP ID
rs7209493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31870304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCCACCAACAGTATATAAGGT[C/T]TACCCTGCAGCCTTGCCAATATCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP6 PubMed Links

Gene Details

Gene
UTP6
Gene Name
UTP6, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018428.2 Intron NP_060898.2
XM_005258012.2 Intron XP_005258069.1
XM_011524997.2 Intron XP_011523299.1
XM_017024845.1 Intron XP_016880334.1

View Full Product Details