Product Details

SNP ID
rs12982834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1288983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCTGTCCTGTTGGACCTGGCC[A/G]GCTGCCCTCACCTGTCCTGGCCACA
Phenotype
MIM: 602756
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf24 PubMed Links

Gene Details

Gene
C19orf24
Gene Name
chromosome 19 open reading frame 24
There are no transcripts associated with this gene.

Gene
EFNA2
Gene Name
ephrin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001405.3 Intron NP_001396.2
XM_017026449.1 Intron XP_016881938.1
XM_017026450.1 Intron XP_016881939.1

View Full Product Details