Product Details

SNP ID
rs6956128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:25224748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCAACCATGTTATTCTGGCATA[C/T]TATTTAATTTCCATCATAATGTTTT
Phenotype
MIM: 616984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPVF PubMed Links

Gene Details

Gene
NPVF
Gene Name
neuropeptide VF precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022150.3 1012 UTR 3 NP_071433.3

View Full Product Details