Product Details

SNP ID
rs10256042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20332818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGATTGGATTGTTGGAAAATTA[C/T]ACAACTAAGTAATAGGACGAATCAT
Phenotype
MIM: 604160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB8 PubMed Links

Gene Details

Gene
ITGB8
Gene Name
integrin subunit beta 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002214.2 Intron NP_002205.1
XM_011515393.2 Intron XP_011513695.1
XM_011515394.2 Intron XP_011513696.1
XM_011515396.1 Intron XP_011513698.1
XM_017012178.1 Intron XP_016867667.1
XM_017012179.1 Intron XP_016867668.1
XM_017012180.1 Intron XP_016867669.1
XM_017012181.1 Intron XP_016867670.1
XM_017012182.1 Intron XP_016867671.1
XM_017012183.1 Intron XP_016867672.1
Gene
LOC101927811
Gene Name
uncharacterized LOC101927811
There are no transcripts associated with this gene.

View Full Product Details