Product Details
- SNP ID
-
rs8071475
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59896559 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACTGGGACTTTTCTTTTCACTTTC[C/T]TGTGTTTCAAGAACAGTAGGTGTTC
- Phenotype
-
MIM: 608938
MIM: 607344
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RPS6KB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80316873] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPS6KB1
- Gene Name
- ribosomal protein S6 kinase B1
- Gene
- TUBD1
- Gene Name
- tubulin delta 1
There are no transcripts associated with this gene.
View Full Product Details