Product Details

SNP ID
rs42395
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132878638 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTTATGATAGGATTTTGATCCA[C/T]TGCCCATTACTACCTTGTGGGAAAA
Phenotype
MIM: 604417 MIM: 611373 MIM: 612080
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AFF4 PubMed Links
Additional Information
For this assay, SNP(s) [rs143672867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AFF4
Gene Name
AF4/FMR2 family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014423.3 6385 UTR 3 NP_055238.1
XM_005271963.4 6385 UTR 3 XP_005272020.1
XM_005271964.4 6385 UTR 3 XP_005272021.1
XM_006714587.3 6385 UTR 3 XP_006714650.1
Gene
LEAP2
Gene Name
liver enriched antimicrobial peptide 2
There are no transcripts associated with this gene.

Gene
UQCRQ
Gene Name
ubiquinol-cytochrome c reductase complex III subunit VII
There are no transcripts associated with this gene.

View Full Product Details