Product Details
- SNP ID
-
rs3744524
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:50383389 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGAGAAAAAATGTCTCCACAGAC[C/T]GAAACAGAAACAACTTTTCCCTTGG
- Phenotype
-
MIM: 610885
MIM: 614854
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EME1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74695075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EME1
- Gene Name
- essential meiotic structure-specific endonuclease 1
There are no transcripts associated with this gene.
- Gene
- LRRC59
- Gene Name
- leucine rich repeat containing 59
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018509.3 |
|
Intron |
|
|
NP_060979.2 |
View Full Product Details