Product Details

SNP ID
rs2711094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:24575341 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAGTCTAGTAGCTCTATATAAGA[C/G]AGAATCTTTTTTTTTCTTTAGGCCA
Phenotype
MIM: 606959
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPP6 PubMed Links

Gene Details

Gene
MPP6
Gene Name
membrane palmitoylated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303037.1 Intron NP_001289966.1
NM_016447.3 Intron NP_057531.2
XM_005249775.4 Intron XP_005249832.1
XM_006715738.3 Intron XP_006715801.1
XM_006715739.3 Intron XP_006715802.1
XM_006715740.3 Intron XP_006715803.1
XM_011515425.2 Intron XP_011513727.1
XM_017012315.1 Intron XP_016867804.1
XM_017012316.1 Intron XP_016867805.1
XM_017012317.1 Intron XP_016867806.1

View Full Product Details