Product Details

SNP ID
hCV2575706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4968438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAACAAATAAATAAGGCAAGTCA[G/T]GAGGGGGCCGAGTCGGGTGCAGGCA
Phenotype
MIM: 611508 MIM: 610056
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CAMTA2 PubMed Links

Gene Details

Gene
CAMTA2
Gene Name
calmodulin binding transcription activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171166.1 4336 UTR 3 NP_001164637.1
NM_001171167.1 4336 UTR 3 NP_001164638.1
NM_001171168.1 4336 UTR 3 NP_001164639.1
NM_015099.3 4336 UTR 3 NP_055914.2
Gene
MIR6864
Gene Name
microRNA 6864
There are no transcripts associated with this gene.

Gene
MIR6865
Gene Name
microRNA 6865
There are no transcripts associated with this gene.

Gene
SPAG7
Gene Name
sperm associated antigen 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004890.2 4336 Intron NP_004881.2

View Full Product Details