Product Details

SNP ID
rs12459628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43729007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGCAGCAGTATATCTAGCTG[A/G]TGTCCACAGTGGCCTGCTGGCAGCA
Phenotype
MIM: 613176
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGC PubMed Links

Gene Details

Gene
IRGC
Gene Name
immunity related GTPase cinema
There are no transcripts associated with this gene.

Gene
SMG9
Gene Name
SMG9, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005259057.2 1631 Silent Mutation XP_005259114.1
XM_011527113.1 1631 Missense Mutation XP_011525415.1
XM_011527114.1 1631 Silent Mutation XP_011525416.1
XM_011527115.1 1631 Silent Mutation XP_011525417.1
XM_011527116.1 1631 Silent Mutation XP_011525418.1
XM_017026988.1 1631 Silent Mutation XP_016882477.1

View Full Product Details