Product Details
- SNP ID
-
rs650314
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:49249685 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCGGGTGCGGCCCCAGGAGATGC[A/C]ATTCATCTGCCAGGGCCACCGGGCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ40194
PubMed Links
Gene Details
- Gene
- FLJ40194
- Gene Name
- uncharacterized FLJ40194
There are no transcripts associated with this gene.
View Full Product Details