Product Details

SNP ID
rs617182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49229912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGTCTCTGGCGCCTCTTGGGA[A/G]AATTGCAGCTGGAGCTCCCCCCTTG
Phenotype
MIM: 606363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABI3 PubMed Links

Gene Details

Gene
ABI3
Gene Name
ABI family member 3
There are no transcripts associated with this gene.

Gene
PHOSPHO1
Gene Name
phosphoethanolamine/phosphocholine phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143804.1 Intron NP_001137276.1
NM_178500.3 Intron NP_848595.1
XM_017024271.1 Intron XP_016879760.1

View Full Product Details