Product Details

SNP ID
rs11769907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39742470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGAGTCCCACGCGCGGTAGTA[A/G]CACACACCGGCCTTGGAGGGCGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00265 PubMed Links

Gene Details

Gene
LINC00265
Gene Name
long intergenic non-protein coding RNA 265
There are no transcripts associated with this gene.

View Full Product Details