Product Details

SNP ID
rs3787031
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:57192197 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCATCATCCTTGGCCTGCAAGCAT[C/T]CTCTGGTGGACCAGGTCTCATTCCA
Phenotype
MIM: 604668
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF264 PubMed Links

Gene Details

Gene
ZNF264
Gene Name
zinc finger protein 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003417.4 Intron NP_003408.1
XM_011527522.2 Intron XP_011525824.1

View Full Product Details