Product Details
- SNP ID
-
rs6510057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57326650 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGCACAGGCTGTCCTTTGTTCAAA[C/G]CAGAGCTGATCTACCAGTTGGATCA
- Phenotype
-
MIM: 616847
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF543
PubMed Links
Gene Details
- Gene
- ZNF543
- Gene Name
- zinc finger protein 543
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213598.3 |
520 |
Missense Mutation |
CCA,GCA |
P55A |
NP_998763.2 |
View Full Product Details