Product Details
- SNP ID
-
rs10221208
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58970524 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTGACTTAAACATTAACTCTTTT[C/T]AAAATGGGCTTGGAGAGATCAGGGT
- Phenotype
-
MIM: 605073
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPM1E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76524595] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPM1E
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1E
- Gene
- TRIM37
- Gene Name
- tripartite motif containing 37
View Full Product Details