Product Details

SNP ID
rs15886
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:69247148 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTATCTTTTCTATGCATTAGTTATG[A/T]ATTAGTTTTTTATTTAGATTGGAAC
Phenotype
MIM: 612508 MIM: 612503
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
There are no transcripts associated with this gene.

Gene
ABCA5
Gene Name
ATP binding cassette subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018672.4 6536 UTR 3 NP_061142.2
NM_172232.3 6536 UTR 3 NP_758424.1

View Full Product Details