Product Details

SNP ID
rs11871637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61412874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCGGGGCTGGGCCCTCGCGTTCT[C/T]TCTGCGGGAAGTGAGCCGCCGGCCT
Phenotype
MIM: 600747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf82 PubMed Links

Gene Details

Gene
C17orf82
Gene Name
chromosome 17 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203425.2 1124 UTR 3 NP_982249.2
Gene
TBX2
Gene Name
T-box 2
There are no transcripts associated with this gene.

View Full Product Details