Product Details

SNP ID
rs34569523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2516622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGGACAGATGGACCCCGTTTT[C/T]GGAGTCCAAGACCTCATTCTCACCC
Phenotype
MIM: 604430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNG7 PubMed Links

Gene Details

Gene
GNG7
Gene Name
G protein subunit gamma 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052847.2 Intron NP_443079.1
XM_017026606.1 Intron XP_016882095.1
Gene
LOC101929097
Gene Name
uncharacterized LOC101929097
There are no transcripts associated with this gene.

View Full Product Details