Product Details

SNP ID
rs2910335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141240223 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGGACCGGGAAGAATTGTGTAGA[A/G]TCACTGAGCCTTGTGTGCTGCAATT
Phenotype
MIM: 606341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB15 PubMed Links

Gene Details

Gene
PCDHB15
Gene Name
protocadherin beta 15
There are no transcripts associated with this gene.

Gene
PCDHB18P
Gene Name
protocadherin beta 18 pseudogene
There are no transcripts associated with this gene.

Gene
PCDHB19P
Gene Name
protocadherin beta 19 pseudogene
There are no transcripts associated with this gene.

View Full Product Details